missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress HOE 32020

MedChemExpress HOE 32020 is a Hoechst stain, which is a blue fluorescent dyes used to stain DNA.

Encompass_Preferred

585.90€ - 3514.05€

Specifica

Content And Storage 4°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light)
Shipping Condition Room temperature
Solubilità DMSO : ≥ 69 mg/mL (155.78 mM)
Peso molecolare 442.94 g/mol
CAS 23554-99-6
Visualizza altre specifiche

Products 4
Product Code Brand Quantità Price Quantity & Availability  
Product Code Brand Quantità Price Quantity & Availability  
16479257
Encompass_Preferred
missing translation for 'viewDocuments'
MedChemExpress
HY-15629
5 mg
585.90€
5mg
View alternative products
missing translation for 'toSaveUpto' 
»
Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!
16436856
Encompass_Preferred
missing translation for 'viewDocuments'
MedChemExpress
HY-15629
100 mg
3514.05€
100mg
View alternative products
missing translation for 'toSaveUpto' 
»
Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!
16422787
Encompass_Preferred
missing translation for 'viewDocuments'
MedChemExpress
HY-15629
50 mg
2511.00€
50mg
View alternative products
missing translation for 'toSaveUpto' 
»
Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!
16406708
Encompass_Preferred
missing translation for 'viewDocuments'
MedChemExpress
HY-15629
10 mg
837.00€
10mg
View alternative products
missing translation for 'toSaveUpto' 
»
Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!
Description

Description

Specifications

Specifications

4°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light)
DMSO : ≥ 69 mg/mL (155.78 mM)
23554-99-6
Room temperature
442.94 g/mol
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
MedChemExpress HOE 32020

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.