missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human ACE2 (aa1-615) Fc Chimera Recombinant Protein
Recombinant Protein
Marca: Invitrogen™ RP87693
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
RP-87677 has been tested in indirect ELISA.
Angiotensin-converting enzyme 2 (ACE2) plays a central role in vascular, renal, and myocardial physiology. In contrast to its homolog ACE, ACE2 expression is restricted to heart, kidney, and testis. Recently. ACE2 has also been shown to be a functional receptor of the SARS coronavirus. The normal function of ACE2 is to convert the inactive vasoconstrictor angiotensin I (AngI) to Ang1-9 and the active form AngII to Ang1-7, unlike ACE, which converts AngI to AngII. While the role of these vasoactive peptides is not well understood, lack of ACE2 expression in ace2-/ace2- mice leads to severely reduced cardiac contractility, indicating its importance in regulating heart function.
Specifica
Q9BYF1 | |
Control, ELISA, Western Blot | |
59272 | |
Human ACE2 (aa1-615) Fc Chimera | |
HEK293 cells | |
2010305L05Rik; ACE 2; Ace2; ACEH; ACE-related carboxypeptidase; angiotensin I converting enzyme (peptidyl-dipeptidase A) 2; angiotensin I converting enzyme 2; angiotensin-converting enzyme 2; Angiotensin-converting enzyme homolog; anigotensin I converting enzyme 2; anigotensin-converting enzyme-related carboxypeptidase; metalloprotease MPROT15; peptidyl-dipeptidase A; Processed angiotensin-converting enzyme 2; renal angiotensin-converting enzyme 2; UNQ868/PRO1885 | |
ACE2 | |
Recombinant | |
Human ACE2 recombinant protein contains amino acids 1 -615 with a human Fc tag | |
HEK293 cells | |
>95% by SDS-PAGE under reducing conditions |
1 mg/mL | |
PBS with no preservative | |
110 kDa | |
1 mg | |
RUO | |
ACE2 | |
Unconjugated | |
Fc-tag | |
-20°Cor to +80°C if preferred | |
Liquid |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Invitrogen™ Human ACE2 (aa1-615) Fc Chimera Recombinant Protein > 1 mg; Unlabeled
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto