Learn More
Gibco™ Human Granzyme B Recombinant Protein, PeproTech®
Recombinant Protein
84.25€ - 5400.00€
Specifica
Numero di accesso | P10144 |
---|---|
Da utilizzare con (applicazione) | Functional Assay |
Formato | Lyophilized |
Formulazione | protein with no preservative |
ID gene (immissione) | 3002 |
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
30019600
|
Gibco™
140-18-500UG |
500 μg |
3240.00€
500µg |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
30019313
|
Gibco™
140-18-2UG |
2 μg |
84.25€
2µg |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
30019435
|
Gibco™
140-18-250UG |
250 μg |
1892.00€
250µg |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
30019479
|
Gibco™
140-18-1MG |
1 mg |
5400.00€
1mg |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
30019980
|
Gibco™
140-18-10UG |
10 μg |
204.00€
10µg |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
Descrizione
Recombinant Human Granzyme B has been confirmed to be a mixture of the pro-peptide and mature forms when tested by N-terminal sequencing. Recombinant Human Granzyme B is a 235 amino acid protein, which includes the mature 227 amino acid sequence, as well as an 8 amino acid C-terminal His-tag. Due to glycosylation, this protein migrates to an approximate molecular weight of 30-40 kDa, under reducing and non-reducing conditions.
Granzyme B is a member of the granzyme serine protease family, and is found in the granules of cytotoxic T cells and NK cells. Granzyme B has been described as CGL1 (cathepsin G-like-1), a serine protease expressed only in cytotoxic T-lymphocytes after cell activation, and CTLA-1 (cytotoxic T lymphocyte-associated serine esterase 1) based on identification of mRNA in various cytotoxic T cells, but not observed in non-cytotoxic lymphoid cells. Granzyme B is crucial for the rapid induction of target cell death by apoptosis, induced by interaction with cytotoxic T cells. The receptor involved in this process has been identified as mannose 6-phosphate receptor which functions as a death receptor for Granzyme B during cytotoxic T cell-induced apoptosis. Granzyme B enters target cells to cleave caspase-3 and initiate the caspase cascade leading to DNA fragmentation and apoptosis. Granzyme B can also act through a mitochondrial apoptosis pathway by cleaving the Bid protein. Granzymes are neutral serine proteases, which are stored in specialized lytic granules of cytotoxic T lymphocytes (CTLs) and in natural killer (NK) cells. A number of granzymes (A to G) have been isolated and cloned from mouse CTLs and NK cells, however in man, fewer have been cloned and identified.Specifica
P10144 | |
Lyophilized | |
3002 | |
Human Granzyme B | |
<1 EU/ μg | |
Granzyme B | |
Unconjugated | |
IIGGHEAKPH SRPYMAYLMI WDQKSLKRCG GFLIRDDFVL TAAHCWGSSI NVTLGAHNIK EQEPTQQFIP VKRPIPHPAY NPKNFSNDIM LLQLERKAKR TRAVQPLRLP SNKAQVKPGQ TCSVAGWGQT APLGKHSHTL QEVKMTVQED RKCESDLRHY YDSTIELCVG DPEIKKTSFK GDSGGPLVCN KVAQGIVSYG RNNGMPPRAC TKVSSFVHWI KKTMKRYHHH HHHHH | |
CHO cells | |
≥ 95% by SDS-PAGE gel and HPLC analyses |
Functional Assay | |
protein with no preservative | |
26.6 kDa | |
RUO | |
AI553453; C11; Cathepsin G-like 1; CCP1; CCP-1/C11; CCPI; CGL1; CGL-1; CSPB; CSP-B; Ctla1; Ctla-1; CTSGL1; cytotoxic cell protease 1; cytotoxic serine protease B; Cytotoxic T lymphocyte associated serine esterase 1; cytotoxic T-lymphocyte proteinase 2; cytotoxic T-lymphocyte-associated serine esterase 1; fragmentin; fragmentin 2; fragmentin-2; GLP I; GLP III; GLP-1; granzyme 2; granzyme B; granzyme B (granzyme 2, cytotoxic T-lymphocyte-associated serine esterase 1); granzyme B(G,H); Granzyme-2; GranzymeB; granzyme-like protein 1; granzyme-like protein I; GRB; GZB; Gzmb; HLP; human lymphocyte protein; Human lymphocyte protein (Hlp); Lymphocyte protease; Natural killer cell protease 1; OTTHUMP00000028189; RNKP-1; SECT; T-cell serine protease 1-3 E | |
Determined by its ability to cleave chromogentic Granzyme B substrate IEPD-pNA | |
Recombinant | |
-20°C | |
Lyophilized |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.