Learn More
PeproTech® Human IL-19, Animal-Free Recombinant Protein
Recombinant Protein
Marca: PeproTech® AF-200-19-10UG
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Recombinant Human IL-19 is a 35.8 kDa homodimer of two 154 amino acid chains. In solution, IL-19 exists predominantly as a non-disulfide-linked dimer.
The protein encoded by this gene is a cytokine that belongs to the IL10 cytokine subfamily. This cytokine is found to be preferentially expressed in monocytes. It can bind the IL20 receptor complex and lead to the activation of the signal transducer and activator of transcription 3 (STAT3). A similar cytokine in mouse is reported to up-regulate the expression of IL6 and TNF-alpha and induce apoptosis, which suggests a role of this cytokine in inflammatory responses. Alternatively spliced transcript variants encoding the distinct isoforms have been described.Specifica
Q9UHD0 | |
Lyophilized | |
35.8 | |
-20°C | |
<1 | |
Determined by its ability to stimulate the proliferation of HEK-Blue TM-IL-19 reporter cells | |
Recombinant | |
purified |
Functional Assay | |
protein with no preservative | |
10 μg | |
RUO | |
IL-19 | |
Unconjugated | |
E. coli |