Learn More
PeproTech® Human IL-19, Animal-Free Recombinant Protein
Recombinant Protein
100.00€ - 5650.00€
Specifica
Numero di accesso Q9UHD0
Da utilizzare con (applicazione) Functional Assay
Formato Lyophilized
Formulazione protein with no preservative
Peso molecolare 35.8
Visualizza altre specifiche
Prodotti 6
Codice del prodotto Marca Quantità Prezzo Quantità e disponibilità  
Codice del prodotto Marca Quantità Prezzo Quantità e disponibilità  
30019401
Vedere i documenti
PeproTech®
AF-200-19-500UG
500 μg
3395.00€
500µg
30019533
Vedere i documenti
PeproTech®
AF-200-19-2UG
2 μg
100.00€
2µg
30019710
Vedere i documenti
PeproTech®
AF-200-19-250UG
250 μg
1979.00€
250µg
30019739
Vedere i documenti
PeproTech®
AF-200-19-1MG
1 mg
5650.00€
1mg
30019286
Vedere i documenti
PeproTech®
AF-200-19-10UG
10 μg
221.00€
10µg
30019308
Vedere i documenti
PeproTech®
AF-200-19-100UG
100 μg
1176.00€
100µg
Descrizione
Descrizione
Recombinant Human IL-19 is a 35.8 kDa homodimer of two 154 amino acid chains. In solution, IL-19 exists predominantly as a non-disulfide-linked dimer.
The protein encoded by this gene is a cytokine that belongs to the IL10 cytokine subfamily. This cytokine is found to be preferentially expressed in monocytes. It can bind the IL20 receptor complex and lead to the activation of the signal transducer and activator of transcription 3 (STAT3). A similar cytokine in mouse is reported to up-regulate the expression of IL6 and TNF-alpha and induce apoptosis, which suggests a role of this cytokine in inflammatory responses. Alternatively spliced transcript variants encoding the distinct isoforms have been described.
Specifica
Specifica
Q9UHD0
Lyophilized
35.8
RUO
IL-19
Unconjugated
E. coli
Functional Assay
protein with no preservative
-20°C
<1
Determined by its ability to stimulate the proliferation of HEK-Blue TM-IL-19 reporter cells
Recombinant
purified