missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Iron-Supplemented Bovine Calf Serum (New Zealand)

A cost-effective alternative to Fetal Bovine Serum (FBS)

Marca:  Cytiva SH30414.03IR

Dettagli aggiuntivi : Peso : 1.15000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16615765

Questo articolo non è attualmente disponibile o è stato dismesso.
La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative.


Descrizione

Descrizione

  • Sourced from New Zealand with complete traceability back to original source
  • Age of animal at time of collection less than 10 days old
  • Filtered through sequential 100 nm pore size-rated filters
  • Virus panel testing according to 9 CFR 113.53
  • Low in antibodies and high in growth factors
  • Supplemented with iron
  • Each serum lot is pooled using true pool technology to ensure uniformity and consistency between bottles
Specifica

Specifica

Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, SGPT, SGOT, pH, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorus, Osmolality, Iron, TIBC, Percent Saturation, Glucose, and IgG
New Zealand
Calf Bovine Serum
Sterile
Irradiated
≤ 25 EU/mL
≤ 25 mg/dL
HyClone™
Store at or below -10°C
Liquid
1000 mL
Sequential-filtered, 100 nm
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
No
Yes
Yes
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali