Learn More
uvex i-5 Welding Safety Glasses
Sfoglia Tutto Uvex Prodotti
Click to view available options
Lens Tint:
Welding Shade 1.7
Welding Shade 3
Welding Shade 5
Descrizione
Sophisticated features ensure superior wearer comfort even during long periods of use.
General features
- Sporty premium welding safety glasses with a variety of features and customization options
- Robust polycarbonate lens with proven uvex infradur coating technology – permanently anti-fog and scratch-resistant lens
- Optional extra frame provides greater protection against particles, dust and dirt
- Metal-free
- Frame color: black, green
Protection features
- Certified in line with EN 166 (personal eye protection) and EN 169 (welding shade)
- IR protection according to EN 169 with perfect color recognition according to EN 172
- UV400 – 100% protection against dangerous UVA and UVB radiation to 400 nm
Comfort features
- X-tended eyeshield: soft-seal frame technology wraps the forehead and provides protection against particle penetration
- X-tended sideshield offers additional side protection
- Ergonomically shaped frame with multi-stage adjustable side arms allows a high degree of customization in order to guarantee a good fit on any head or face shape
- Soft ear pieces for a comfortable, secure hold without creating pressure points
- Soft, flexible nose pad adapts to the wearer and ensures a comfortable, pressure-free fit
Specifica
Specifica
Tipo di lente | Infradur Plus |
Lens Coating | Anti-fog |
Descrizione | Anti-fog, Resistant to Welding Sparks, Scratch Resistant |
Certifications/Compliance (Europe) | EN 166, EN 169 |
Lens Tint | Welding Shade 5 |
Product Type | Welding Spectacles |
Colore telaio | Green, Black |
Materiale | Polycarbonate |
Monouso | Reusable |
Lens Material | Policarbonato |
Vedi altri risultati |
Offerte speciali
-
Diversi produttori
Gear Up for Safety
Save up to 60% on gloves, apparel, and moreCodice sconto: 24131Scade: 2025-06-08
nuova offerta Shop Now
Titolo del prodotto
uvex i-5 Welding Safety Glasses >