Learn More
Idoxuridine, MedChemExpress
MedChemExpress Idoxuridine (5-Iodo-2′-deoxyuridine, 5-IUdR, IdUrd) is an iodinated thymidine analogue that competitively inhibits phosphorylases. Idoxuridine can inhibit viral activity, particularly viral eye infections, including herpes simplex keratitis, by inhibiting DNA polymerase and affecting viral replication. Idoxuridine against feline herpesvirus has the IC50 value of 4.3 μM. Idoxuridine shows anti-orthopoxvirus activity.
Marca: Medchem Express HY-B0307
Descrizione
Idoxuridine (5-Iodo-2′-deoxyuridine, 5-IUdR, IdUrd) is an iodinated thymidine analogue that competitively inhibits phosphorylases. Idoxuridine can inhibit viral activity, particularly viral eye infections, including herpes simplex keratitis, by inhibiting DNA polymerase and affecting viral replication. Idoxuridine against feline herpesvirus has the IC50 value of 4.3 μM. Idoxuridine shows anti-orthopoxvirus activity.Specifica
Idoxuridine | |
White | |
C9H11IN2O5 | |
5-Iodo-2′-deoxyuridine 5-IUdR IdUrd | |
OC[C@@H]1[C@H](C[C@H](N2C(NC(C(I)=C2)=O)=O)O1)O | |
354.1 | |
Research | |
COVID-19-anti-virus |
54-42-2 | |
Research | |
1 g | |
DMSO : 125 mg/mL (353.01 mM; Need ultrasonic) | |
354.1 | |
98.0% | |
Powder |
Il programma Encompass di Fisher Scientific offre articoli che non fanno parte del nostro portafoglio di distribuzione. Questi prodotti in genere non hanno immagini o descrizioni dettagliate. Tuttavia, ci impegniamo a migliorare la vostra esperienza di acquisto. Si prega di utilizzare il modulo sottostante per fornire un feedback relativo al contenuto di questo prodotto.
Research purposes only