Learn More
IL-17RA/IL-17R Antibody (CL10309), Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP3-21175-100ul
593.60 EUR valido fino al 2025-03-28
Usa il codice promo "25306" per avere il tuo prezzo promozionale.
Descrizione
IL-17RA/IL-17R Monoclonal antibody specifically detects IL-17RA/IL-17R in Human samples. It is validated for Western Blot
Specifica
IL-17RA/IL-17R | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.2, 40% glycerol | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
CL10309 | |
Western Blot 1 ug/ml | |
CD217, CD217 antigen, CDw217interleukin 17 receptor, hIL-17R, IL-17 receptor A, IL-17RAMGC10262, IL17Rinterleukin-17 receptor A, interleukin 17 receptor A | |
This antibody was developed against a Synthetic Peptide of Q96F46. Actual immunogen sequence is proprietary. | |
100 μg | |
Innate Immunity | |
23765 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.