Learn More
DWK Life Sciences Kimble™ Clear Screw Thread Sample Vials without Closures
Packaged in corrugated trays with partitions
Marca: DWK Life Sciences 608122385
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Manufactured from clear 33 expansion borosilicate glass
Specifica
Clear | |
Borosilicate Glass | |
Screw | |
85 mm | |
20-400 |
23 mm | |
24 mL | |
6.5 dr. | |
ASTM Type I, Class A |