Learn More
DWK Life Sciences Kimble™ Kontes™ Replacement Tube For All-Glass Dounce Tissue Grinders
A range of replacement tubes for Dounce Tissue Grinders, from 2mL to 100mL.
Marca: DWK Life Sciences 885303-0002
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
A range of replacement tubes for Dounce Tissue Grinders, from 2mL to 100mL.
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifica
Case of 1 | |
2 mL | |
Dounce all Glass tissue grinders |
0.06 oz. | |
Tissue Grinder Tube | |
0.06 oz., 2mL |