Learn More
DWK Life Sciences Kimble™ Open-Top PolypropyleneScrew Thread Closures with Bonded White PTFE Liners
Intended for single-use injection
Marca: DWK Life Sciences 73814BK13425
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Manufactured using the highest quality raw materials
- Designed for an ideal fit with Kimble Chase autosampler vials
- Pre-assembled caps and septa minimizes contamination due to handling
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifica
PTFE | |
Polypropylene | |
13-425 |
Black | |
Autoclavable |