Learn More
DWK Life Sciences Kimble™ Kontes™ ULTRA-WARE™ Heavy-Wall Filter Flasks
Heavy-wall borosilicate glass provides mechanical strength for vacuum filtration
Marca:  DWK Life Sciences 953760-2002
Dettagli aggiuntivi : Peso : 0.80000kg
Codice prodotto. 11752754
  • 466.00€ / Pezzo
Spedizione stimata: 17-10-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
Heavy-wall borosilicate glass provides mechanical strength for vacuum filtration
Kimble Chase KimCote™ plastic-coated glassware provides an added measure of safety in the event of breakage
  • KimCote™ plastic safety coating
  • ≠2 side arm for connection to 0..25 in. (6mm) I.D. flexible tubing
  • 125mL flask: With No. 5 stopper joint
  • 1, 2 and 4L flasks: With No. 8 stopper joint
Specifica
Specifica
0.6 cm
2000 mL
Borosilicate Glass
8