Learn More
Tocris Bioscience™ L-755,507
Very potent and selective β3 partial agonist
Marca: Tocris Bioscience™ 2197/50
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
L-755,507 is a potent β3-adrenergic receptor partial agonist > 1000-fold selective over β1- and β2-adrenoceptors (EC50 values are 0.43, 580 and > 10000 nM for activation of cloned human β3-, β1- and β2-adrenoceptors respectively). Stimulates lipolysis in rhesus adipocytes in vitro (EC50 = 3.9 nM). Enhances CRISPR-mediated homology-directed repair (HDR) efficiency 2-3-fold for large fragments and ∽9-fold for point mutations, in human induced pluripotent stem cells (iPSCs).Specifica
>98% | |
L-755,507 | |
L-755,507 |
50 mg | |
584.73 |
Titolo del prodotto
For Research Use Only