Learn More
Kappa Light Chain Ab-1 Mouse Monoclonal Antibody, Epredia™
Recommended for Immunohistochemistry (Formalin/paraffin), Epredia™ Kappa Light Chain Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.
Marca: Epredia MS-143-R7
229.68 EUR valido fino al 2024-12-31
Usa il codice promo "21628" per avere il tuo prezzo promozionale.
Descrizione
Antibody to the κ light chain of immunoglobulin is reportedly useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkin's lymphomas. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.Host Species: Mouse
Clone: L1C1
Isotype: IgG1/k
Species Reactivity: Human. Does not react with rat. Others not known.
Epitope: Not determined
Immunogen: B-lymphoma cells
Molecular Weight: 25kDa
Positive Control: Tonsil
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifica
Kappa Light Chain Ab-1 | |
Monoclonal | |
Unconjugated | |
B lymphoma cells | |
IVD | |
Primary | |
Prediluted |
Immunohistochemistry (Paraffin) | |
L1C1 | |
Mouse | |
7 mL | |
Hematopoietic | |
Human | |
IgG2a κ |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.