Learn More
Labconco™ SpillStopper™ Work Surface for 4 Ft. Protector™Premier™Hoods
Labconco SpillStopper™ Work Surface is dished and contoured to conform to the interior liner to contain spills.
Marca: Labconco™ 9500410
Dettagli aggiuntivi : Peso : 54.43000kg
Descrizione
- Corrosion-resistant epoxy resins molded
- Front edge with large radius to aerodynamically direct airflow into hood
- Pre-drilled holes for venting and plumbing pass-throughs provided
- Some surfaces with cutouts for surface-mounted service fixture and/or 6 x 3 in. (15.2 x 7.6cm) cup sinkcup sink
- Flat work surfaces may also be coupled with adjustable height base stand to provide auxiliary workspace
Order Info
Service fixture and cup sink sold separately
Specifica
Left Rear | |
Epoxy Resin |