Learn More
Labconco™ SpillStopper™ Work Surface for 4 Ft. Protector™Premier™Hoods
Labconco SpillStopper™ Work Surface is dished and contoured to conform to the interior liner to contain spills.
Marca:  Labconco™ 9500401
Dettagli aggiuntivi : Peso : 10.50000kg
 vedi altre versioni di questo prodotto
Codice prodotto. 17621037
  • 1650.00€ / Pezzo
Spedizione stimata: 03-10-2024
per visualizzare lo stock disponibile
Descrizione
Descrizione
  • Corrosion-resistant epoxy resins molded
  • Front edge with large radius to aerodynamically direct airflow into hood
  • Pre-drilled holes for venting and plumbing pass-throughs provided
  • Some surfaces with cutouts for surface-mounted service fixture and/or 6 x 3 in. (15.2 x 7.6cm) cup sinkcup sink
  • Flat work surfaces may also be coupled with adjustable height base stand to provide auxiliary workspace
Order Info
Service fixture and cup sink sold separately
Specifica
Specifica
Right Rear
Epoxy Resin