Learn More
Smeg™ Laboratory Glassware Washer
Clean a wide range of glassware with this washer. Smeg™ Laboratory Glassware Washers are manufactured with high quality materials and feature faultless, intuitive performance. Specific accessories ensure high quality washing of even the most difficult and dirtier instruments.
Marca: Smeg™ GW0160S1
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Microprocessor electronic control
- Active thermodynamic drying
- Alkaline powder detergent dispenser
- Peristaltic pump for acid liquid neutralizer
- Washing chamber and internal door in AISI 304 stainless steel
- External panels made in AISI 304 stainless steel
- 2 independent washing levels through telescopic guides
- Incorporated water softener with salt sensor
- Cold water and demineralized water connections
- Electronic door lock system for user safety
- Without trolleys
Specifica
Stainless Steel | |
7 default programs | |
490 x 525 x 570mm |
Glassware Washer | |
Washing up to 85°C | |
1/N/PE 230V - 50Hz - 2,8 kW max |