Learn More
LPCAT2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA596301
338.68 EUR valido fino al 2025-03-29
Usa il codice promo "24111" per avere il tuo prezzo promozionale.
Allerta:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Descrizione
Immunogen sequence: LPSMVSRNEN AQVPLIGRLL RAVQPVLVSR VDPDSRKNTI NEIIKRTTSG GEWPQILVFP EGTCTNRSCL ITFKPGAFIP GVPVQPVLLR YPNKLDTVTW TWQGYTFIQL CMLTFCQLFT KVEVEFMPVQ VPNDEEKNDP VLFANKVRNL MAEALGIPVT DHTYEDCRLM ISAGQLTLPM EAGLVEFTKI SRKLKLDWDG VRKHLDEYAS IASSSKGGRI GIEEFAKYLK LPVSDVLRQL FALFDRNHDG SIDFREYVIG LAVLCNPSNT EEIIQVAFKL FDVDEDGYIT EEEFSTILQA SLGVPDLDVS GLFKEIAQGD SISYEEFKSF ALKHPEYAKI FTTYLDLQTC HVFSLPKEVQ TTPSTASNKV SPEKHEESTS DKKDD; Positive Samples: HeLa, Jurkat; Cellular Location: Endoplasmic reticulum membrane, Golgi apparatus membrane, Lipid droplet, Single-pass type II membrane protein
This gene encodes a member of the lysophospholipid acyltransferase family. The encoded enzyme may function in two ways: to catalyze the biosynthesis of platelet-activating factor (1-O-alkyl-2-acetyl-sn-glycero-3-phosphocholine) from 1-O-alkyl-sn-glycero-3-phosphocholine, and to catalyze the synthesis of glycerophospholipid precursors from arachidonyl-CoA and lysophosphatidylcholine. The encoded protein may function in membrane biogenesis and production of platelet-activating factor in inflammatory cells. The enzyme may localize to the endoplasmic reticulum and the Golgi. [provided by RefSeq, Feb 2009].
Specifica
LPCAT2 | |
Polyclonal | |
Unconjugated | |
LPCAT2 | |
1-acylglycerol-3-phosphate O-acyltransferase 11, 1-acylglycerophosphocholine O-acyltransferase, 1-AGP acyltransferase 11, 1-AGPAT 11, 1-alkylglycerophosphocholine O-acetyltransferase, A330042H22, Acetyl-CoA:lyso-PAF acetyltransferase, acetyl-CoA:lyso-platelet-activating factor acetyltransferase, acyl-CoA:lysophosphatidylcholine acyltransferase 2, acyl-CoA:lysophosphatidylcholine acyltransferase member 1, acyltransferase like 1, acyltransferase like 1A, acyltransferase-like 1, acyltransferase-like 1-A, AGPAT11, AYTL1, Aytl1a, LPAAT-alpha, lpafat1, LPC acyltransferase 2, Lpcat2, LPCAT-2, Lpcat2a, lyso-PAF acetyltransferase, LysoPAFAT, lysoPAFAT/LPCAT2, lysoPC acyltransferase 2, lysophosphatidic acid acyltransferase alpha, Lysophosphatidylcholine acyltransferase 2, lyso-platelet-activating factor (PAF) acetyltransferase, RGD1563994 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
54947 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
2.63 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide, pH 7.3 | |
Q7L5N7 | |
LPCAT2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 160-544 of human LPCAT2 (NP_0603092) | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.