Learn More
Value Plastics Male Luer Integral Lock Ring to Female Luer Thread Style Coupler
Prevent leaks with integral lock ring mechanism
Marca: Value Plastics 50120-04
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Luer fittings allow for a simple yet secure twist-lock connection. Easily transition from a male luer to female luer fitting. Ideal for general laboratory and single-use applicationsSpecifica
ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP | |
Male Luer Integral Lock Ring, Female Luer Thread | |
Polypropylene |
Titolo del prodotto
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.