Learn More
Euro Protection™ Marble High-Cut Safety Shoes
Euro Protection™ Marble High-Cut Safety Shoes have a padded collar and bellows tongue for a snug fit and include a composite toe cap for protection.
Marca: Euro Protection™ 9MARH42
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Padded collar and bellows tongue for snug fit.
- Footbridge supplying torsional rigidity.
- Nubuck leather upper.
- Moisture wicking non-woven lining.
- Removable, textile and foam insole.
- Composite toe cap.
- Non-metallic (composite fabric) puncture resistant sole.
- Injected polyurethane midsole.
- Polyurethane outsole.
- A spare pair of shoelaces.
Specifica
Brown | |
Leather | |
Removable, textile and foam insole, non-metallic (composite fabric) puncture-resistant sole, injected polyurethane midsole, polyurethane outsole. | |
Composite | |
EN ISO 20345 : 2011 |
Moisture wicking non-woven lining. | |
42 | |
Polyurethane, Composite Fabric | |
694 g per shoe in size 8 | |
Safety Shoe |