Learn More
Greiner Bio-One MASTERBLOCK™ 96 Deep Well Round Bottom 1mL Storage Plate
Suitable for cultivating bacteria or yeasts. Greiner Bio-One MASTERBLOCK™ 96 Deep Well Round Bottom 1mL Storage Plate is also suitable for storing sample material.
Marca: Greiner Bio-One 780261
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Free of detectable DNase, RNase, human DNA
- Non-pyrogenic
- Manufactured under DIN ISO 9001 guidelines
- Sealable with adhesive films and heat sealer
- High temperature resistance
- High chemical resistance
- A
Specifica
Standard | |
8 x 12 | |
Polypropylene | |
Sterile | |
1 mL | |
1 | |
Chimney |
Without Lid | |
Natural | |
96 | |
Non-treated | |
Round Bottom (U-well) | |
50/Cs. |