Learn More
MED15 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca:  Proteintech CL488-11566-20UL
Dettagli aggiuntivi : Peso : 0.00100kg
Codice prodotto. 18088985
  • 166.92€ / 20µL
Spedizione stimata: 02-07-2024
per visualizzare lo stock disponibile
Descrizione
Descrizione
The MED15 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human MED15. This antibody recognizes human antigen. The MED15 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifica
Specifica
MED15
1000 μg/mL
ARC105, CAG7A, CTG repeat protein 7a, CTG7A, DKFZp686A2214, DKFZp762B1216, FLJ42282, FLJ42935, MED15, mediator complex subunit 15, PCQAP, TIG 1, TIG1, TNRC7, TPA inducible gene 1 protein
20 μL
51586
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Liquid
Immunofluorescence
CoraLite Plus 488
Rabbit
Primary
Human
150 kDa