Learn More
MED15 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech CL488-11566-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The MED15 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human MED15. This antibody recognizes human antigen. The MED15 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifica
MED15 | |
1000 μg/mL | |
ARC105, CAG7A, CTG repeat protein 7a, CTG7A, DKFZp686A2214, DKFZp762B1216, FLJ42282, FLJ42935, MED15, mediator complex subunit 15, PCQAP, TIG 1, TIG1, TNRC7, TPA inducible gene 1 protein | |
20 μL | |
51586 | |
Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
Liquid |
Immunofluorescence | |
CoraLite Plus 488 | |
Rabbit | |
Primary | |
Human | |
150 kDa | |
IgG |