missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(Methoxycarbonylsulfamoyl)triethylammonium hydroxide, 97%, inner salt, Thermo Scientific Chemicals
47.70€ - 757.00€
Identificatori chimici
CAS | 29684-56-8 |
---|---|
Formula molecolare | C8H18N2O4S |
Molecular Weight (g/mol) | 238.30 |
Numero MDL | MFCD00077815 |
InChI Key | YSHOWEKUVWPFNR-UHFFFAOYSA-N |
Sinonimo | methoxycarbonylsulfamoyl triethylammonium hydroxide, burgess reagent, carbomethoxysulfamoyltriethylammonium hydroxide, methoxycarbonylsulfamoyltriethylammonium hydroxide, methoxycarbonylsulphamoyltriethylammonium hydroxide, methoxy-carbonylsulfamoyltriethylammonium hydroxide, methoxycarbonylsulfa-moyl-triethylammonium hydroxide, methoxycarbonylsulphamoyl-triethylammonium hydroxide |
PubChem CID | 11032497 |
SMILES | CC[N+](CC)(CC)S(=O)(=O)[N-]C(=O)OC |
Codice del prodotto | Marca | Quantità | Imballaggio | Prezzo | Quantità e disponibilità | ||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantità | Imballaggio | Prezzo | Quantità e disponibilità | ||||
10571331
|
Thermo Scientific Acros
294510010 |
1 g | Glass bottle |
47.70€
1g |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | ||||
10615723
|
Thermo Scientific Acros
294510050 |
5 g | Glass bottle |
177.00€
5g |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | ||||
11925741
|
Thermo Scientific Acros
294510250 |
25 g | Glass bottle |
757.00€
25g |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | ||||
Descrizione
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identificatori chimici
29684-56-8 | |
238.30 | |
YSHOWEKUVWPFNR-UHFFFAOYSA-N | |
11032497 |
C8H18N2O4S | |
MFCD00077815 | |
methoxycarbonylsulfamoyl triethylammonium hydroxide, burgess reagent, carbomethoxysulfamoyltriethylammonium hydroxide, methoxycarbonylsulfamoyltriethylammonium hydroxide, methoxycarbonylsulphamoyltriethylammonium hydroxide, methoxy-carbonylsulfamoyltriethylammonium hydroxide, methoxycarbonylsulfa-moyl-triethylammonium hydroxide, methoxycarbonylsulphamoyl-triethylammonium hydroxide | |
CC[N+](CC)(CC)S(=O)(=O)[N-]C(=O)OC |
Specifica
(Methoxycarbonylsulfamoyl)triethylammonium hydroxide | |
76.0°C to 79.0°C | |
Authentic | |
C8H18N2O4S | |
MFCD00077815 | |
15,1505 | |
YSHOWEKUVWPFNR-UHFFFAOYSA-N | |
triethyl(methoxycarbonylsulfamoyl)azanium;hydroxide | |
11032497 | |
97% |
29684-56-8 | |
Yellow | |
96% min | |
CH3O2CNSO2N(C2H5)3 | |
14,208 | |
methoxycarbonylsulfamoyl triethylammonium hydroxide, burgess reagent, carbomethoxysulfamoyltriethylammonium hydroxide, methoxycarbonylsulfamoyltriethylammonium hydroxide, methoxycarbonylsulphamoyltriethylammonium hydroxide, methoxy-carbonylsulfamoyltriethylammonium hydroxide, methoxycarbonylsulfa-moyl-triethylammonium hydroxide, methoxycarbonylsulphamoyl-triethylammonium hydroxide | |
CC[N+](CC)(CC)S(=O)(=O)[N-]C(=O)OC | |
238.30 | |
238.31 | |
Crystalline Powder |
Sicurezza e movimentazione
GHS Signal Word: Warning
Identificativo prodotto
Avvertenza
Categoria di pericolo
Indicazioni di pericolo
Consigli di prudenza
- (Methoxycarbonylsulfamoyl)triethylammonium hydroxide, inner salt
Avvertenza
- Attenzione
Categoria di pericolo
- Lesione oculare grave/irritazione oculare Categoria 2
- Corrosione/irritazione della pelle Categoria 2
- Tossicità specifica per organi bersaglio (esposizione singola) Categoria 3
Indicazioni di pericolo
- H319-Provoca grave irritazione oculare.
- H335-Può irritare le vie respiratorie.
- H315-Provoca irritazione cutanea.
Consigli di prudenza
- P261-Evitare di respirare la polvere/i fumi/i gas/la nebbia/i vapori/gli aerosol.
- P280-Indossare guanti/indumenti protettivi/Proteggere gli occhi/il viso.
- P302+P352-IN CASO DI CONTATTO CON LA PELLE: lavare abbondantemente con acqua/.
- P305+P351+P338-IN CASO DI CONTATTO CON GLI OCCHI: sciacquare accuratamente per parecchi minuti. Togliere le eventuali lenti a contatto se è agevole farlo. Continuare a sciacquare.
RUO – Research Use Only