Learn More
Thermo Scientific Chemicals Methylene Blue, high purity, biological stain
22.80€ - 260.00€
Identificatori chimici
CAS | 122965-43-9 |
---|---|
Formula molecolare | C16H18ClN3S |
Molecular Weight (g/mol) | 319.85 |
Numero MDL | MFCD00150006 |
InChI Key | CXKWCBBOMKCUKX-UHFFFAOYSA-M |
Sinonimo | Basic Blue 9; C.I. 52015 |
PubChem CID | 16211647 |
IUPAC Name | [7-(dimethylamino)phenothiazin-3-ylidene]-dimethylazanium;chloride;hydrate |
SMILES | [Cl-].CN(C)C1=CC=C2N=C3C=CC(C=C3SC2=C1)=[N+](C)C |
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
11443697
|
Thermo Scientific Chemicals
A18174.14 |
25 g |
22.80€
25g |
In stock
Eseguire il login per visualizzare lo stock disponibile |
|||||
11453697
|
Thermo Scientific Chemicals
A18174.22 |
100 g |
61.60€
100g |
In stock
Eseguire il login per visualizzare lo stock disponibile |
|||||
11463697
|
Thermo Scientific Chemicals
A18174.36 |
500 g |
260.00€
500g |
In stock
Eseguire il login per visualizzare lo stock disponibile |
|||||
Descrizione
Methylene blue is used as a dye in different staining procedures viz. Wright's stain and Jenner's stain. It also serves as an indicator and medicine. Further, it is used to examine RNA and DNA under the microscope or in a gel. It is widely used as a redox indicator in analytical chemistry. It acts as a photosensitizer utilized to generate singlet oxygen in order to make organic peroxides by a Diels-Alder reaction. It is also used in the sulfide analysis. It plays an important role in aquaculture to treat fungal infections.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethylene blue is used as a dye in different staining procedures viz. Wright′s stain and Jenner′s stain. It also serves as an indicator and medicine. Further, it is used to examine RNA and DNA under the microscope or in a gel. It is widely used as a redox indicator in analytical chemistry. It acts as a photosensitizer utilized to generate singlet oxygen in order to make organic peroxides by a Diels-Alder reaction. It is also used in the sulfide analysis. It plays an important role in aquaculture to treat fungal infections.
Solubility
Soluble in water, alcohol, acetic acid, glycerol and chloroform. Insoluble in ether.
Notes
Incompatible with strong oxidizing agents.
Identificatori chimici
122965-43-9 | |
319.85 | |
CXKWCBBOMKCUKX-UHFFFAOYSA-M | |
16211647 | |
[Cl-].CN(C)C1=CC=C2N=C3C=CC(C=C3SC2=C1)=[N+](C)C |
C16H18ClN3S | |
MFCD00150006 | |
Basic Blue 9; C.I. 52015 | |
[7-(dimethylamino)phenothiazin-3-ylidene]-dimethylazanium;chloride;hydrate |
Specifica
Methylene Blue | |
biological stain | |
High purity | |
MFCD00150006 | |
14,6060 | |
Soluble in water,alcohol,acetic acid,glycerol and chloroform. Insoluble in ether. | |
[Cl-].CN(C)C1=CC=C2N=C3C=CC(C=C3SC2=C1)=[N+](C)C | |
319.85 | |
319.86 (Anhydrous) |
∼180°C (decomposition) | |
122965-43-9 | |
C16H18ClN3S | |
3599847 | |
Basic Blue 9; C.I. 52015 | |
CXKWCBBOMKCUKX-UHFFFAOYSA-M | |
[7-(dimethylamino)phenothiazin-3-ylidene]-dimethylazanium;chloride;hydrate | |
16211647 |
Sicurezza e movimentazione
GHS H Statement
H302
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
missing translation for 'einecsNumber' : 200-515-2
missing translation for 'rtecsNumber' : SO5600000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Methylene Blue, high purity, biological stain
Avvertenza
- Attenzione
Categoria di pericolo
- Tossicità acuta Categoria 4
Indicazioni di pericolo
- H302-Nocivo se ingerito.
Consigli di prudenza
- P301+P312-IN CASO DI INGESTIONE accompagnata da malessere: contattare un CENTRO ANTIVELENI/medico/
Informazioni supplementari
- MIXTURE LIST-Contiene: Methylene Blue hydrate
RUO – Research Use Only