Learn More
Metro™ Stainless Laboratory Worktable Accessory, Wire Shelf
Expand your workspace in your lab. Metro Stainless Laboratory Worktable shelves come in a choice of stainless steel or epoxy-coated.
Marca:  Metro™ LT48WS
Dettagli aggiuntivi : Peso : 3.63200kg
Codice prodotto. 15937041
  • 336.00€ / Pezzo
Spedizione stimata: 07-02-2025
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
  • Shelves mount on three-sided frames with use of clips
  • Load rated for a maximum of 150 lbs.
  • Also compatible with MetroMax i Lab Worktables
Specifica
Specifica
40.3 in.
Wire Shelf
68 kg
102.5 cm
Stainless
4.5 kg
10 lb.
Metro™ MetroMax i™ and other Stainless Steel worktables
3-Sided Frame
150 lb.