Learn More
Thermo Scientific™ Microcentrifuga MicroCL 21
Applicazioni di ricerca di alto livello che richiedono prestazioni di microcentrifugazione fino a 21,100xg.
Marca: Thermo Scientific™ 75002549
Descrizione
Risparmio di tempo con la rapida lavorazione dei campioni delle microcentrifughe Thermo Scientific MicroCL 21, che offrono produttività, versatilità, sicurezza e convenienza in uno strumento da laboratorio compatto e facile da utilizzare. I leggeri rotori per microcentrifuga sono studiati per garantire un'accelerazione fino a 21,100 xg in appena 12 secondi.
Features:
- Exceptional acceleration of up to 21,000xg in as little as 12 seconds
- Choose from 7 optional microcentrifuge rotors to find the ideal fit for your application
- Outstanding corrosion resistance with lightweight engineered polymer rotors
- Standard rotor runs 24 microcentrifuge tubes in a single row, from 1.5 to 2.0 mL tubes to mini-preps and spin columns
- Ergonomic one-click rotor opening and closing, as well as transparent design for added safety and convenience with Thermo Scientific ClickSeal biocontainment lids
- Eliminate the need for adapters with unique dual row rotor
- Simplify operation with intuitive controls, easy-to-read displays and fast one-click centrifuge lid closure
- Sleek, lightweight, space-saving design
- Durable and easy to clean and maintain
- Quiet performance promotes a safe, stress-free environment
- Conforms to the latest clinical and safety standards, including 98/79/EC In Vitro Diagnostic Directive
Specifica
24 x 1.5/2 mL | |
21,100 x g | |
Ambient temperature | |
Benchtop Centrifuge | |
MicroCL 21 Centrifuge, Ventilated | |
2 Years | |
1 to 99 min. (1 min. increments); HOLD | |
8.9 x 9.6 x 13.8 in. (22.5 x 24.3 x 35.2 cm) | |
Thermo Scientific | |
230 V |
14,800 rpm | |
56 dB(A) | |
CSA, CE, IVD | |
Dual Row 18 x 2.0/0.5mL rotor with screw-on lid | |
230 V 50/60 Hz | |
No | |
IEC 61010-1, IEC 61010-2-020 (refer to manual for more details) | |
23.1lb. (10.5 kg) | |
50/60 Hz | |
Dual Row 18 x 2/0.5 mL Rotor with Screw-On Lid |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.