Learn More
Cytiva (formerly Pall Lab) MicroFunnel™ ST Filters Funnel with GN-6 Membrane
Microfunnel ST filter funnels are available in gamma-irradiated, double-bagged packaging saving valuable time during entry into cleanrooms and hoods.
Marca: Cytiva (formerly Pall Lab) 4811
Dettagli aggiuntivi : Peso : 1.86200kg
Descrizione
- For research use only
Specifica
100 mL | |
Sterile | |
GN-6 Metricel Mixed Cellulose Ester |
0.45 μm | |
Filtering Funnel |