Learn More
KARTELL Microtitration Plate
The Microtitration Plate has 96 sample wells, available in two shapes: flat bottom and V-bottom.
Marca: KARTELL 0262200
34.21 EUR valido fino al 2024-12-31
Usa il codice promo "21898" per avere il tuo prezzo promozionale.
Dettagli aggiuntivi : Peso : 0.25000kg
Descrizione
- Multiple-well plates with 96 sample wells in clear PS.
- Flat bottom or V-bottom wells, single wrapped sterile.
Specifica
No Barcode | |
8 x 12 | |
None | |
Polystyrene | |
96 | |
Non-treated | |
Certified Quality |
Standard | |
Not Autoclavable | |
Clear | |
0.281 mL | |
Non-sterile | |
Conical Bottom (V-well) |