Learn More
3M™ Molecular Detection Assay 2
Used with the 3M™ Molecular Detection System for the rapid and specific detection of shiga toxin-producing e.coli species in enriched food and environmental samples.
Marca: 3M™ 7100194012
Dettagli aggiuntivi : Peso : 0.01100kg
Descrizione
- One assay protocol and fewer steps than most tests reduces repetitive tasks and makes it easy to train employees.
- Ready-to-use and pre-dispensed reagents provide convenience and minimize the chance of human error.
- Color-coded assay tubes by organism and matching software screens allow for better tracking and easy identification.
- The ability to run all assays simultaneously and the flexibility to test up to 96 samples in each run helps you operate more efficiently.
- Real-time amplification and detection process identifies positive results in as early as 15 minutes and allows you to make critical decisions faster.
Specifica
STEC Gene Screen | |
Microbiology Media | |
Food and Beverage, Environmental Test | |
96 Pack |