Learn More
3M™ Molecular Detection Assay 2
Used with the 3M™ Molecular Detection System for the rapid and specific detection of shiga toxin-producing e.coli species in enriched food and environmental samples.
513.04€ - 956.12€
Specifica
Descrizione | STEC Gene Screen |
---|---|
Product Type | Microbiology Media |
Test Type | Food and Beverage, Environmental Test |
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
17218550
|
3M™
7100194012 |
96 Pack |
956.12€
1 set |
Spedizione stimata: 11-12-2024
Eseguire il login per visualizzare lo stock disponibile |
|||||
17228550
|
3M™
7100194040 |
48 Pack |
513.04€
1 set |
Spedizione stimata: 11-12-2024
Eseguire il login per visualizzare lo stock disponibile |
|||||
Descrizione
- One assay protocol and fewer steps than most tests reduces repetitive tasks and makes it easy to train employees.
- Ready-to-use and pre-dispensed reagents provide convenience and minimize the chance of human error.
- Color-coded assay tubes by organism and matching software screens allow for better tracking and easy identification.
- The ability to run all assays simultaneously and the flexibility to test up to 96 samples in each run helps you operate more efficiently.
- Real-time amplification and detection process identifies positive results in as early as 15 minutes and allows you to make critical decisions faster.
Specifica
STEC Gene Screen | |
Microbiology Media | |
Food and Beverage, Environmental Test |