missing translation for 'onlineSavingsMsg'
Learn More
N-Fmoc-S-trityl-L-cysteine, 95%, Thermo Scientific Chemicals Codice prodotto.: 11307139

N-Fmoc-S-trityl-L-cysteine, 95%, Thermo Scientific Chemicals

Codice prodotto. 11307139
25 g
Click to view available options
Quantità:
1 g
5 g
25 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Codice prodotto. 11307139

Marca: Thermo Scientific Alfa Aesar H27363.14

Richiesta di formato massivo o personalizzato

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

It is potentially useful for proteomics studies, and solid phase peptide synthesis techniques. Cysteine is versatile amino acid involved with many biological processes, including the formation of disulfide bonds - a critical component of protein structure. This compound could be useful as an unusual amino acid analog to aid in the deconvolution of protein structure and function.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is potentially useful for proteomics studies, and solid phase peptide synthesis techniques. Cysteine is versatile amino acid involved with many biological processes, including the formation of disulfide bonds - a critical component of protein structure. This compound could be useful as an unusual amino acid analog to aid in the deconvolution of protein structure and function.

Solubility
Insoluble in water. Soluble in most organic solvents.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 103213-32-7
Formula molecolare C37H30NO4S
Molecular Weight (g/mol) 584.71
Numero MDL MFCD00038538
InChI Key KLBPUVPNPAJWHZ-UMSFTDKQSA-M
Sinonimo fmoc-cys trt-oh, fmoc-s-trityl-l-cysteine, fmoc-l-cys trt-oh, n-fmoc-s-trityl-l-cysteine, fmoc-cys trt, n alpha-fluorenylmethyloxycarbonyl-s-tritylcysteine, r-9h-fluoren-9-yl methyl 1-hydroxy-3-tritylthio propan-2-yl carbamate, fmoc-s-trityl-cys, n-9h-fluoren-9-ylmethoxy carbonyl-s-triphenylmethyl-l-cysteine, l-cysteine, n-9h-fluoren-9-ylmethoxy carbonyl-s-triphenylmethyl
PubChem CID 128239
IUPAC Name (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-tritylsulfanylpropanoic acid
SMILES [O-]C(=O)[C@H](CSC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifica

Materiale o nome chimico N-Fmoc-S-trityl-L-cysteine
Punto di fusione 171°C to 173°C
Quantità 25 g
Informazioni di solubilità Insoluble in water. Soluble in most organic solvents.
Optical Rotation +17° (c=1 in THF)
Peso formulazione 585.71
Percent Purity 95%

Certificati

Per poter ottenere i certificati è necessario indicare un numero di lotto. Per individuare il numero di lotto su ordini precedenti usi l'area dedicata allo stato degli ordini.

5 Risultati trovati
Numero di lotto Tipo di certificato Data Catalog Number
Numero di lotto10243443 Tipo di certificatoCertificato di analisi Data28/10/2022 Catalog NumberH27363.14,H27363.06
Numero di lotto10237400 Tipo di certificatoCertificato di analisi Data12/04/2022 Catalog NumberH27363.14,H27363.06
Numero di lotto10180583 Tipo di certificatoCertificato di analisi Data07/12/2021 Catalog NumberH27363.14,H27363.06
Numero di lotto10228715 Tipo di certificatoCertificato di analisi Data07/12/2021 Catalog NumberH27363.14,H27363.06
Numero di lotto10171886 Tipo di certificatoCertificato di analisi Data07/12/2021 Catalog NumberH27363.14,H27363.06

RUO – Research Use Only

Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto
N-Fmoc-S-trityl-L-cysteine, 95%, Thermo Scientific Chemicals >

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato