Learn More
Thermo Scientific™ Nalgene™ Color-Coded LDPE Wash Bottles
Simplify content identification with these narrow-mouth wash bottles that feature colored closures with an integrated stem.
42.00€
Specifica
Capacità (sistema metrico) | 500 ml |
---|---|
Capacità (sistema imperiale) | 16 oz. |
Colore | Incolore |
Descrizione | Con stelo e tubo di prelievo |
Materiale | LDPE |
Codice del prodotto | Marca | Colore chiusura | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Colore chiusura | Prezzo | Quantità e disponibilità | |||||
10055280
|
Thermo Scientific™
2422-2500 |
Giallo |
42.00€
pezzi 6 |
In stock
Eseguire il login per visualizzare lo stock disponibile |
|||||
10732191
|
Thermo Scientific™
2422-3500 |
Blue |
42.00€
pezzi 6 |
In stock
Eseguire il login per visualizzare lo stock disponibile |
|||||
10780442
|
Thermo Scientific™
2422-5500 |
Rosso |
42.00€
pezzi 6 |
In stock
Eseguire il login per visualizzare lo stock disponibile |
|||||
10517131
|
Thermo Scientific™
2422-4500 |
Green |
42.00€
pezzi 6 |
In stock
Eseguire il login per visualizzare lo stock disponibile |
|||||
Descrizione
High quality squeeze bottles with color-coded closures for easy identification. Thermo Scientific™ Nalgene™ Color-Coded LDPE Wash Bottles are made from lab quality low density polyethylene (LDPE) for dependably low leachables and chemical compatability with a wide range of lab reagents. Enjoy smooth, precision, sputter-free fluid dispensing with an ergonomic soft squeeze. Ideal for rinsing tubes and lab apparatus, bringing volumetric flasks to final volume and other precision rinsing and solution dispensing needs. The dispensing nozzle is molded in one piece with the bottle closure eliminating risk of separation when the bottle is squeezed. Cut back tip to increase flow.
- Color-coded top allows you to coordinate reagents with industry storage codes.
- Tip can be cut back to provide increased flow.
- Tight fit of draw tube and closure prevents separation when squeezing bottle.
- Flexible LDPE bottle provides soft, ergonomic squeeze.
- Sputter-free, precision stream.
- Bottle and Carton are readily recyclable in most communities.
- Manufactured from non-toxic materials.
.
Specifica
500 ml | |
Incolore | |
LDPE | |
6 | |
Not Autoclavable | |
Narrow | |
224 mm | |
8.8 in. | |
28 mm | |
A vite |
16 oz. | |
Con stelo e tubo di prelievo | |
24 | |
Yes | |
No | |
Cylindrical | |
Color-coded Wash Bottle | |
Polipropilene | |
72.9 mm | |
Yes |