Learn More
Thermo Scientific™ Raccordi a Y per tubi Nalgene™
Maximize liquid flow with non-tapered hose barbed Y-Type Connectors, molded of polypropylene resin, free of animal derived components.
Marca:  Thermo Scientific™ 6152-0187
136.80 EUR valido fino al 2024-08-02
Usa il codice promo "21702" per avere il tuo prezzo promozionale.
Dettagli aggiuntivi : Peso : 0.06200kg
Codice prodotto. 11704743
  • 144.00€ / pezzi 72
Spedizione stimata: 21-08-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
Maximize liquid flow with non-tapered hose barbed Thermo Scientific™ Nalgene™ Tubing Y-Type Connectors, molded of polypropylene resin, free of animal derived components. Products are double-bagged to maintain cleanliness.
  • Polypropylene construction
  • Autoclavable
Specifica
Specifica
Tubing Connector
Autoclavable
4.76 mm
3/16 in. I.D.
Connettore tipo Y
3/16 in.
Polipropilene
12/Pk., 72/Cs.
Offerte speciali
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.