Learn More
Thermo Scientific™ National Polypropylene and Glastic™ Vials with 8-425 Finish
Thermo Scientific™ National Polypropylene and Glastic Vials provide an economical alternative to glass for some applications.
Marca: Thermo Scientific™ C4013-15GST
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Thermo Scientific™ National Polypropylene and Glastic Vials provide an economical alternative to glass for some applications.
- Precision formed tips are designed to aid sample recovery for limited-volume applications
- Glastic vial features a glass insert pre-inserted inside a clear TPX vial
Specifica
150µL Glastic Glass Insert/ TPX MicroVials | |
Glass, TPX | |
<2 μL | |
100/Pk. |
Clear | |
450 μL | |
300 μL | |
8-425 |