Learn More
Thermo Scientific™ National Polypropylene and Glastic™ Vials with 8-425 Finish
Thermo Scientific™ National Polypropylene and Glastic Vials provide an economical alternative to glass for some applications.
22.54€ - 199.00€
Descrizione
Thermo Scientific™ National Polypropylene and Glastic Vials provide an economical alternative to glass for some applications.
- Precision formed tips are designed to aid sample recovery for limited-volume applications
- Glastic vial features a glass insert pre-inserted inside a clear TPX vial
Specifica
Clear | |
8-425 |