Learn More
Motic™ Objective, CCIS Plan Achromatic EC-H PL for BA410
Color-corrected infinity system objective using multi-layer coated glass for enhanced contrast. Motic™ Objective, CCIS Plan Achromatic EC-H PL for BA410 is designed for field curvature, spherical aberration, and chromatic aberration correction.
Marca: Motic™ 1101001702891
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Suitable for BA410 Series Upright Compound Microscope.
- Color-corrected infinity system.
- Enhanced contrast. Field curvature correction.
- Chromatic aberration correction for 2 colors.
- Spherical correction for 1 color.
Specifica
Objective | |
NA: 0.80/S, WD: 0.35mm | |
BA410E Microscopes | |
60X |