missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ OncoStat Panel, SNAP-ChIP™ Spike-in

SNAP-ChIP (Sample Normalization and Antibody Profiling Chromatin ImmunoPrecipitation) uses DNA-barcoded semi-synthetic nucleosomes bearing distinct histone mutations as spike-in ChIP controls.

Marca:  Invitrogen™ A47343

 vedi altre versioni di questo prodotto

Codice prodotto. 17168855

  • 435.00€ / 10 reazioni
Spedizione stimata: 27-01-2025
per visualizzare lo stock disponibile
Esplora altre offerte speciali
Aggiungi al carrello
Questo articolo non è restituibile. Consulta la politica di reso

Descrizione

Descrizione

  • SNAP-ChIP (Sample Normalization and Antibody Profiling Chromatin ImmunoPrecipitation) uses DNA-barcoded semi-synthetic nucleosomes bearing distinct histone mutations as spike-in ChIP controls.
  • Homogenous, fully defined standards faithfully represent target mononucleosomes in the experimental sample
  • Nucleosomes are subjected to rigorous quality control for lot-to-lot consistency
  • Unique DNA barcodes can be distinguished from experimental sample genomes
  • Spike-ins provide a direct readout of antibody performance in ChIP
  • Ability to monitor technical variability between samples
  • Defined standards enable universal normalization across experiments
  • SNAP-ChIP panels are directly compatible with your current ChIP workflow, with semi-synthetic nucleosomes bearing the mutation of interest immunoprecipitated and processed alongside sample chromatin.
  • Users can directly monitor antibody capability (specificity and enrichment) in a ChIP experiment by measuring the recovery of on- and off-target nucleosomes. The SNAP spike-ins can be further used as a defined standard to normalize your ChIP data, thereby controlling for technical variability across experiments.
  • The OncoStat Panel consists of a pool of nucleosomes harboring seven well-studied histone H3.3 mutations that have been implicated in cancer (histone H3.3K4M, H3.3K9M, H3.3K27M, H3.3G34R, H3.3G34V, H3.3G34W and H3.3K36M) plus a wild-type H3.3 control. Further, each of these eight nucleosomes is wrapped by two independent DNA barcodes (16 total barcodes in the panel), allowing for an internal technical replicate in each ChIP reaction.
  • A single spike-in of the panel allows users to check antibody specificity by examining the post-IP recovery of on- versus off-target SNAP-ChIP nucleosomes, supporting the generation of high quality ChIP data.
TRUSTED_SUSTAINABILITY
Specifica

Specifica

Store at -5 to -30°C.
10 Reactions
OncoStat™, SNAP-ChIP™
Recombinant Nucleosome Panel
10 reactions
SNAP-ChIP Panel
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali

Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato

For Research Use Only. Not for use in diagnostic procedures.