missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OncoStat Panel, SNAP-ChIP™ Spike-in
SNAP-ChIP (Sample Normalization and Antibody Profiling Chromatin ImmunoPrecipitation) uses DNA-barcoded semi-synthetic nucleosomes bearing distinct histone mutations as spike-in ChIP controls.
435.00€ - 3645.00€
Specifica
Content And Storage | Store at -5 to -30°C. |
---|---|
Descrizione | Recombinant Nucleosome Panel |
Product Line | OncoStat™, SNAP-ChIP™ |
Product Type | SNAP-ChIP Panel |
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
17168855
|
Invitrogen™
A47343 |
10 reactions |
435.00€
10 reazioni |
Spedizione stimata: 27-01-2025
Eseguire il login per visualizzare lo stock disponibile |
|||||
17183239
|
Invitrogen™
A47355 |
100 reactions |
3645.00€
100 reazioni |
Spedizione stimata: 27-01-2025
Eseguire il login per visualizzare lo stock disponibile |
|||||
Descrizione
- SNAP-ChIP (Sample Normalization and Antibody Profiling Chromatin ImmunoPrecipitation) uses DNA-barcoded semi-synthetic nucleosomes bearing distinct histone mutations as spike-in ChIP controls.
- Homogenous, fully defined standards faithfully represent target mononucleosomes in the experimental sample
- Nucleosomes are subjected to rigorous quality control for lot-to-lot consistency
- Unique DNA barcodes can be distinguished from experimental sample genomes
- Spike-ins provide a direct readout of antibody performance in ChIP
- Ability to monitor technical variability between samples
- Defined standards enable universal normalization across experiments
- SNAP-ChIP panels are directly compatible with your current ChIP workflow, with semi-synthetic nucleosomes bearing the mutation of interest immunoprecipitated and processed alongside sample chromatin.
- Users can directly monitor antibody capability (specificity and enrichment) in a ChIP experiment by measuring the recovery of on- and off-target nucleosomes. The SNAP spike-ins can be further used as a defined standard to normalize your ChIP data, thereby controlling for technical variability across experiments.
- The OncoStat Panel consists of a pool of nucleosomes harboring seven well-studied histone H3.3 mutations that have been implicated in cancer (histone H3.3K4M, H3.3K9M, H3.3K27M, H3.3G34R, H3.3G34V, H3.3G34W and H3.3K36M) plus a wild-type H3.3 control. Further, each of these eight nucleosomes is wrapped by two independent DNA barcodes (16 total barcodes in the panel), allowing for an internal technical replicate in each ChIP reaction.
- A single spike-in of the panel allows users to check antibody specificity by examining the post-IP recovery of on- versus off-target SNAP-ChIP nucleosomes, supporting the generation of high quality ChIP data.
Specifica
Store at -5 to -30°C. | |
Recombinant Nucleosome Panel | |
OncoStat™, SNAP-ChIP™ | |
SNAP-ChIP Panel |
For Research Use Only. Not for use in diagnostic procedures.
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto