Learn More
Kolmi Hopen™ Op-Air™ One Type IIR Medical Mask with Visor
The built-in visor of this medical mask helps protect the wearer from being splashed by biological fluids.
Marca:  Kolmi Hopen™ M34301V-30
Dettagli aggiuntivi : Peso : 1.33000kg
Codice prodotto. 17740209
  • 35.00€ / pezzi 25
Spedizione stimata: 29-08-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
  • Protection against human liquids, bacteria, particles, and viruses
  • Anatomical folding reduces exposure to face
  • Inner layer designed to wick away moisture and sweat
  • Transparent polyester visor with anti-fog treatment on both sides
Specifica
Specifica
Honey
Polypropylene
(EU) 2017/745, EN 14683:2019+AC:2019, ISO 9001, ISO 13485
Medical
25 Pk.
Ties
Pleated with Eyeshield