Learn More
Kolmi Hopen™ Op-Air™ Type II Medical Masks with Earloops
Pleated medical mask with anti-fog nose clip can be used during transport of contagious patients.
Marca: Kolmi Hopen™ M14321-30
5.56 EUR valido fino al 2024-09-30
Usa il codice promo "21675" per avere il tuo prezzo promozionale.
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- High filtration media (polypropylene)
- Two wide and comfortable elastic bands
- Very soft polypropylene edges
- Anatomical folding limits exposure to face
- Integrated anti-fog nose bar
- Fiberglass-free filter
- Produced in ISO 8 clean room (particle and microbiological control)
Specifica
Green | |
Polypropylene | |
CE, (EU) 2017/745, EN 14683:2019, AC:2019 | |
Medical |
Earloops | |
Pleated | |
Single-use | |
60 Pk. |