Learn More
Thermo Scientific™ Orion Star T900 Series Titrator Accessories
Minimize maintenance downtime with easy to install Orion Star T900 series titrator replacement parts. Orion Star T900 series titrator accessories expand functionality.
Marca: Thermo Scientific™ START-DVK
39.93 EUR valido fino al 2025-04-30
Usa il codice promo "24119" per avere il tuo prezzo promozionale.
Descrizione
Get accessories and replacement parts for the Orion Star T910 pH titrator, Orion Star T920 redox titrator, Orion Star T930 ion titrator and Orion Star T940 all-in-one titrator. These are designed to expand titrator functionality and minimize maintenance downtime.
•Select interchangable burette sizes of 10mL, 20mL or 50mL to customize your titrator
•Bottle holder helps secure titrant to prevent spills and offers flexible placement
•Color-coded tubing and connectors simplify tubing installation and maintenance
•Computer software is designed to make data transfer simple and reliable
Specifica
Orion Star T900 series dispense verification kit | |
Titrator accessory, dispenser | |
Orion Star T910 pH titrator, Orion Star T920 redox titrator, Orion Star T930 ion titrator, Orion Star T940 all-in-one titrator |
Offerte speciali
-
Diversi produttori
The Academic Best Buy Book 2025
Browse exclusive prices for our academic customersCodice sconto: 25081Scade: 2025-12-31
- Diversi produttori