Learn More
Thermo Scientific™ Owl™ EasyCast™ B2 and B3 Mini Gel Electrophoresis System Combs
Eliminate comb assembly with this assortment of heavy-duty, one-piece combs.
Marca: Thermo Scientific™
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
Eliminate comb assembly with Thermo Scientific™ Owl™ EasyCast™ B2 and B3 Mini Gel Electrophoresis System Combs. These heavy-duty, one-piece combs are for use with the EasyCast™ B2 and B3 Mini Gel Electrophoresis Systems.
Heavy-duty, one-piece combs eliminate comb assembly.
Ordering Information:
Cat. Nos. B2-RL-9C, -9D, -25C and -25D are 8 and 12-channel pipette format (1X) and (2X) combs
Customized combs are available (Order SKU # XCM). Please contact your Customer Service or Sales Representative for further details.
Specifica
Rapid-load comb | |
Mini | |
Comb | |
1 mm |
Owl™ EasyCast B2 and B3 Mini Gel Electrophoresis System | |
Owl, EasyCast | |
25 (2X) |