missing translation for 'onlineSavingsMsg'
Learn More
p-Phenylenediamine, 97%, Thermo Scientific Chemicals Codice prodotto.: 11427627

p-Phenylenediamine, 97%, Thermo Scientific Chemicals

Codice prodotto. 11427627
1000 g
Click to view available options
Quantità:
250 g
1000 g
5000 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Codice prodotto. 11427627

Marca: Thermo Scientific Alfa Aesar A15680.0B

Richiesta di formato massivo o personalizzato

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

p-Phenylenediamine is used as a precursor to aramid plastics, Kevlar fibers and to urethane polymers. It serves as a developing agent in photography, antioxidants in rubber products and as a vulcanization accelerator. It is used as a henna surrogate for temporary tattoos and as a histological stain for lipids such as myelin. It is an anti-fade reagent used in fluorescence microscopy to retard photobleaching of fluorescein and other fluorescent dyes. Furthermore, it is also an ingredient in hair dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Phenylenediamine is used as a precursor to aramid plastics, Kevlar fibers and to urethane polymers. It serves as a developing agent in photography, antioxidants in rubber products and as a vulcanization accelerator. It is used as a henna surrogate for temporary tattoos and as a histological stain for lipids such as myelin. It is an anti-fade reagent used in fluorescence microscopy to retard photobleaching of fluorescein and other fluorescent dyes. Furthermore, it is also an ingredient in hair dyes.

Solubility
Soluble in alcohol, chloroform, ether and hot benzene.

Notes
Store in cool place. Incompatible with acids, acid chlorides, acid anhydrides, chloroformates and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 106-50-3
Formula molecolare C6H8N2
Molecular Weight (g/mol) 108.144
Numero MDL MFCD00007901
InChI Key CBCKQZAAMUWICA-UHFFFAOYSA-N
Sinonimo p-phenylenediamine, 1,4-benzenediamine, 1,4-diaminobenzene, 1,4-phenylenediamine, 4-aminoaniline, para-phenylenediamine, p-aminoaniline, p-diaminobenzene, orsin, 4-phenylenediamine
PubChem CID 7814
ChEBI CHEBI:51403
IUPAC Name benzene-1,4-diamine
SMILES C1=CC(=CC=C1N)N

Specifica

Materiale o nome chimico p-Phenylenediamine
Punto di fusione 138°C to 142°C
Punti di ebollizione 267°C to 271°C
Punto d'infiammabilità 156°C (312°F)
Intervallo percentuale saggio 97%
Odore Weak
Quantità 1000 g
Numero UN UN1673
Beilstein 742029
Indice di Merck 14,7285
Informazioni di solubilità Soluble in alcohol,chloroform,ether and hot benzene.
Peso formulazione 108.14
Percent Purity 97%
Vedi altri risultati Mostra meno risultati
compliance-icons compliance-icons
Identificativo prodotto
  • p-Phenylenediamine
Avvertenza
  • Pericolo
Categoria di pericolo
  • Tossicità acuta Categoria 3
  • PERICOLO A LUNGO TERMINE PER L'AMBIENTE ACQUATICO Cronico 1
  • PERICOLO ACUTO PER L'AMBIENTE ACQUATICO Acuto 1
  • Lesione oculare grave/irritazione oculare Categoria 2
  • Sensibilizzazione della pelle Categoria 1
Indicazioni di pericolo
  • H301-Tossico se ingerito.
  • H311-Tossico per contatto con la pelle.
  • H317-Può provocare una reazione allergica cutanea.
  • H319-Provoca grave irritazione oculare.
  • H331-Tossico se inalato.
  • H400-Molto tossico per gli organismi acquatici.
  • H410-Molto tossico per gli organismi acquatici con effetti di lunga durata.
Consigli di prudenza
  • P273-Non disperdere nell'ambiente.
  • P280-Indossare guanti/indumenti protettivi/Proteggere gli occhi/il viso.
  • P302+P352-IN CASO DI CONTATTO CON LA PELLE: lavare abbondantemente con acqua/.
  • P304+P340-IN CASO DI INALAZIONE: trasportare l'infortunato all'aria aperta e mantenerlo a riposo in posizione che favorisca la respirazione.
  • P305+P351+P338-IN CASO DI CONTATTO CON GLI OCCHI: sciacquare accuratamente per parecchi minuti. Togliere le eventuali lenti a contatto se è agevole farlo. Continuare a sciacquare.
  • P312-Contattare un CENTRO ANTIVELENI/un medico/in caso di malessere.
Informazioni supplementari
  • MIXTURE LIST-Contiene: 1,4-Benzenediamine

RUO – Research Use Only

Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto
p-Phenylenediamine, 97%, Thermo Scientific Chemicals >

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato