Learn More
DiscoverX PathHunter™ CHO-K1 mCCR8 β-Arrestin Cell Line
The PathHunter™ CHO-K1 mCCR8 β-Arrestin Cell Line measures mCCR8 (GPCR) activity via recruitment of β-Arrestin2. This cell line ships as two vials of cryopreserved cells. Mouse protein.
Marca: DiscoverX 93-0556C2
Descrizione
The PathHunter™ mCCR8 β-Arrestin cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. CCL1) based activation of mCCR8 GPCR activity via detection of β-arrestin recruitment. This product uses EFC technology (www.discoverx.com/technologies-platforms/enzyme-fragment-complementation-technology). The included cell line overexpresses PK-tagged mCCR8 and EA-tagged β-Arrestin 2. Activation of mCCR8 stimulates the recruitment of β-Arrestin 2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. Please note that this product expresses the Mouse version of this protein.Specifica
β-Arrestin recruitment | |
Cell Lines | |
2 Vials | |
NM_007720.2 |
CHO-K1 | |
CCL1 (murine) | |
mCCR8 |
Il programma Encompass di Fisher Scientific offre articoli che non fanno parte del nostro portafoglio di distribuzione. Questi prodotti in genere non hanno immagini o descrizioni dettagliate. Tuttavia, ci impegniamo a migliorare la vostra esperienza di acquisto. Si prega di utilizzare il modulo sottostante per fornire un feedback relativo al contenuto di questo prodotto.