missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGM2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11022-1-AP-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
EWSR1 is a multifunctional protein that is involved in various cellular processes, including gene expression, cell signaling, and RNA processing and transport. The protein includes an N-terminal transcriptional activation domain and a C-terminal RNA-binding domain.Specifica
PGM2 | |
Polyclonal | |
Unconjugated | |
PGM2 | |
Glucose phosphomutase 2, MSTP006, PGM 2, PGM2, Phosphodeoxyribomutase, phosphoglucomutase 2, Phosphopentomutase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55276 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q96G03 | |
PGM2 | |
PGM2 Fusion Protein Ag1488 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |