Learn More
Proteintech Phantom Dye UV 450 Viability Dye
Phantom Dye UV 450 is a fixable viability dye which reacts with protein amine groups in both dead and live cells.
Marca: Proteintech PD00003-100TESTS
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Due to the compromised cell membrane in dead cells the dye can bind to many more proteins inside the cell, resulting in much higher fluorescence and easy differentiation of Live/Dead cells.
- Phantom Dye UV 450 is a water soluble UV fluorescent dye excited by the UV laser (355 nm) making it an excellent addition to your multicolor panel.
Specifica
Flow Cytometry | |
450 Emission | |
100 Tests |
Titolo del prodotto
Proteintech Phantom Dye UV 450 Viability Dye > 100 Tests