Learn More
BD Phoenix™ ID Broth
Used to make a heavy suspension of a selected colony for processing in the Phoenix™ automated identification and susceptibility testing system. BD Phoenix™ ID Broth is packaged in 100 pre-measured bottles of prepared solution.
Marca: BD 246001
Allerta:
prepared, ready to use, bottle, premeasured, automated, identification, susceptibility, testing, heavy, suspension, innoculate
Descrizione
BD Phoenix™ Automated Identification and Susceptibility Testing System
- Rapid, accurate and reliable detection of known and emerging antimicrobial resistance.
- Workflow efficiency utilizing automated nephelometry resulting in a standardized isolate inoculum and a reduction of technologist labor.
- State-of-the-art data management to monitor, analyze and communicate actionable results directly to laboratorians and clinicians.
Specifica
Microbiology Media | |
Phoenix™ automated identification and susceptibility testing system | |
100Pack |
Preparing a heavy suspension of a colony | |
Pre-filled bottles |
Titolo del prodotto
BD Phoenix™ ID Broth > Phoenix ID Broth: 100Pack