Learn More
Thermo Scientific™ Pierce™ Alkylating Reagents
Alkylating reagents provided in single-use, pre-weighed formats to streamline protein preparation workflows.
102.00€ - 265.00€
Codice del prodotto | Marca | Include | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Include | Prezzo | Quantità e disponibilità | |||||
16336562
|
Thermo Scientific™
A39270 |
Chloroacetamide, No-Weigh™ Format |
102.00€
pezzi 10 |
In stock
Eseguire il login per visualizzare lo stock disponibile |
|||||
15940677
|
Thermo Scientific™
A39271 |
Iodoacetamide, No-Weigh™ Format |
265.00€
pezzi 30 |
In stock Eseguire il login per visualizzare lo stock disponibile |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | ||||
Descrizione
Thermo Scientific Pierce Iodoacetamide or Chloroacetamide are provided in a no-weigh format, where small amounts of reagent are pre-weighed and packaged for single use. This single-use format prevents the contamination and loss of reagent reactivity over time associated with repetitive opening and closing of a single vial. The format enables use of a fresh vial of reagent each time, and eliminates the hassle of weighing out small amounts of reagent.
Alkylating reagents iodoacetamide and chloracetamide are sulfhydryl-reactive alkylating reagents used to block reduced cysteine residues for protein characterization and peptide mapping. Alkylation with iodoacetamide or chloracetamide after cysteine reduction results in the covalent addition of a carbamidomethyl group (57.07 Da) and prevents the formation of disulfide bonds. Reducing agents added after alkylation will react with excess iodoacetamide.Specifica
Pierce™ | |
Alkylating Reagent |